1-Bromo-4-fluorobenzene ≥99%

Synonyms: 4-Bromofluorobenzene

B67201-100G B67201-500G B67201-25G
220111-625EA 64.62 USD
220111-625 220111-627 220111-626
1-Bromo-4-fluorobenzene ≥99%
1-Bromo-4-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1.4946 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00000342
CAS Number: 460-00-4
EINECS: 207-300-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR