1-Bromo-2-fluorobenzene ≥99%

Synonyms: 2-Bromofluorobenzene

B66809-100G B66809-25G
220111-614EA 147.54 USD
220111-614 220111-615
1-Bromo-2-fluorobenzene ≥99%
1-Bromo-2-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 155 °C (1013 hPa)
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000282
CAS Number: 1072-85-1
EINECS: 214-018-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR