N-(2-Bromoethyl)phthalimide

Synonyms: 2-(2-Bromoethyl)isoindoline-1,3-dione

B66302-100G B66302-25G
220112-098EA 99.62 USD
220112-098 220112-099
N-(2-Bromoethyl)phthalimide
N-(2-Bromoethyl)phthalimide
Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Boiling Pt: 318 °C (1013 hPa)
Melting Pt: 79…82 °C
MDL Number: MFCD00005902
CAS Number: 574-98-1
EINECS: 209-379-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR