Phenethyl bromide ≥98%

Warning

Synonyms: (2-Bromoethyl)benzene, 2-Phenylethyl bromide

B65780-500G B65780-5G B65780-100G
220112-091EA 105.54 USD
220112-091 220112-092 220112-090
Phenethyl bromide ≥98%
Phenethyl bromide
Formula: BrCH₂CH₂C₆H₅
MW: 185.06 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –56 °C
Density: 1.37 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000240
CAS Number: 103-63-9
EINECS: 203-130-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR