2-Bromethylamine hydrobromide

Warning

B65705-100G B65705-25G
220112-088EA 107.69 USD
220112-088 220112-089
2-Bromethylamine hydrobromide
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR