α-Phenylbenzyl bromide ≥95%

Danger

Synonyms: Bromodiphenylmethane, α-Phenylbenzyl bromide, Benzhydryl bromide

B65403-100G B65403-25G
220112-073EA 124.46 USD
220112-073 220112-074
α-Phenylbenzyl bromide ≥95%
α-Phenylbenzyl bromide
Formula: (C₆H₅)₂CHBr
MW: 247.13 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 36 - 39 °C
Density: 1.491 g/cm³ (13 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000134
CAS Number: 776-74-9
EINECS: 212-279-8
UN: 1770
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR