Benzamidine hydrochloride hydrate ≥99%

Warning

B6506-5G B6506-100G B6506-25G
220111-356EA 46.11 USD
220111-356 220111-354 220111-355
Benzamidine hydrochloride hydrate ≥99%
Benzamidine hydrochloride hydrate
Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
MW: 174.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR