1-Bromo-3-chloropropane ≥99%

B62404-1KG B62404-250G
220112-063EA 138.92 USD
220112-063 220112-064
1-Bromo-3-chloropropane ≥99%
1-Bromo-3-chloropropane
Formula: BrH₂CCH₂CH₂Cl
MW: 157.44 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: -58 °C
Density: 1.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000998
CAS Number: 109-70-6
EINECS: 203-697-1
UN: 2688
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR