1-Bromo-4-chlorobutane ≥99%

B60800-100G B60800-25G
220112-044EA 154 USD
220112-044 220112-045
1-Bromo-4-chlorobutane ≥99%
1-Bromo-4-chlorobutane
Formula: BrH₂C(CH₂)₂CH₂Cl
MW: 171.46 g/mol
Boiling Pt: 80 °C (1013 hPa)
Density: 1.4895 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001010
CAS Number: 6940-78-9
EINECS: 230-089-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR