1-Bromo-4-chlorobenzene ≥99%

B60428-100G B60428-1KG B60428-500G
220111-511EA 36.62 USD
220111-511 220111-512 220111-513
1-Bromo-4-chlorobenzene ≥99%
1-Bromo-4-chlorobenzene
Formula: C₆H₄BrCl
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 66…68 °C
Density: 1.576 g/cm³ (71 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR