1-Bromo-2-chlorobenzene ≥99%

Warning

B60401-100G B60401-10G
220111-509EA 99.72 USD
220111-509 220111-510
1-Bromo-2-chlorobenzene ≥99%
1-Bromo-2-chlorobenzene
Formula: C₆H₄BrCl
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: –12.1 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR