(±)-α-Bromo-γ-butyrolactone

B59608-100G B59608-25G
220111-264EA 359.69 USD
220111-264 220111-265
(±)-α-Bromo-γ-butyrolactone
(±)-α-Bromo-γ-butyrolactone
Formula: C₄H₅BrO₂
MW: 164.99 g/mol
Boiling Pt: 137…139 °C (8 hPa)
Density: 1.994 g/cm³ (20 °C)
MDL Number: MFCD00005387
CAS Number: 5061-21-2
EINECS: 225-764-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR