3-Bromo-α,α,α-trifluorotoluene ≥99%

Synonyms: 1-Bromo-3-(trifluoromethyl)benzene, 3-Bromobenzotrifluoride

B59004-100G B59004-250G
220112-025EA 114.15 USD
220112-025 220112-026
3-Bromo-α,α,α-trifluorotoluene ≥99%
3-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: 37.5 - 39 °C
Density: 1.6276 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000380
CAS Number: 401-78-5
EINECS: 206-932-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR