Sodium butyrate ≥98.5%

Synonyms: Butyric acid sodium salt, Sodium Butyrate

B5887-5G B5887-250MG B5887-1G
220112-019EA 621.69 USD
220112-019 220112-018 220112-017
Sodium butyrate ≥98.5%
Sodium butyrate
Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: 244 °C
Density: 1.324 g/cm³ (30 °C)
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now

Specification Test Results

Assay 98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR