2-Bromo-α,α,α-trifluorotoluene ≥99%

Danger

Synonyms: 1-Bromo-2-(trifluoromethyl)benzene, 2-Bromobenzotrifluoride

B58806-25G B58806-100G
220112-016EA 31.23 USD
220112-016 220112-015
2-Bromo-α,α,α-trifluorotoluene ≥99%
2-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 167 °C (1013 hPa)
Density: 1.651 g/cm³ (30 °C)
MDL Number: MFCD00000373
CAS Number: 392-83-6
EINECS: 206-879-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR