Bromocresol purple

Synonyms: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

B5880-5G B5880-100G B5880-25G
220112-014EA 42.85 USD
220112-014 220111-946 220112-013
Bromocresol purple
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Melting Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR