4-Bromobenzophenone ≥98%

Synonyms: (4-Bromophenyl)(phenyl)methanone, p-Bromobenzophenone

B58601-100G B58601-25G
220111-943EA 68.92 USD
220111-943 220111-944
4-Bromobenzophenone ≥98%
4-Bromobenzophenone
Formula: C₁₃H₉BrO
MW: 261.12 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 78…82 °C
MDL Number: MFCD00000103
CAS Number: 90-90-4
EINECS: 202-024-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR