3-Bromobenzaldehyde ≥97%

Warning

B57206-100G B57206-25G
220111-274EA 106.08 USD
220111-274 220111-275
3-Bromobenzaldehyde ≥97%
3-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 18…21 °C
Density: 1.587 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00003345
CAS Number: 3132-99-8
EINECS: 221-526-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR