2-Bromobenzaldehyde ≥98%

Warning

Synonyms: o-Bromobenzaldehyde

B57001-25G B57001-10G
220111-936EA 77.75 USD
220111-936 220111-935
2-Bromobenzaldehyde ≥98%
2-Bromobenzaldehyde
Formula: C₇H₅BrO
MW: 185.02 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003300
CAS Number: 6630-33-7
EINECS: 229-622-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR