9-Bromoanthracene ≥94%

B56609-25G B56609-5G
220111-925EA 357.54 USD
220111-925 220111-926
9-Bromoanthracene ≥94%
9-Bromoanthracene
Formula: C₁₄H₉Br
MW: 257.13 g/mol
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR