4-Bromoanisole ≥99%

B56501-100G B56501-50G B56501-500G
220111-919EA 55.46 USD
220111-919 220111-921 220111-920
4-Bromoanisole ≥99%
4-Bromoanisole
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.457 g/cm³ (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000097
CAS Number: 104-92-7
EINECS: 203-252-1
Merck Index: 13,01412

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR