3-Bromoanisole ≥98%

Synonyms: 1-Bromo-3-methoxybenzene

B56498-100G B56498-25G
220111-916EA 134.62 USD
220111-916 220111-917
3-Bromoanisole ≥98%
3-Bromoanisole
Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR