2-Bromoaniline ≥98%

B56420-5G B56420-25G
220111-913EA 37.69 USD
220111-913 220111-912
2-Bromoaniline ≥98%
2-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 27…31 °C
Density: 1.86 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007632
CAS Number: 615-36-1
EINECS: 210-421-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR