Bromoacetyl bromide ≥98%

B56412-100G B56412-5G B56412-500G
220111-909EA 70.97 USD
220111-909 220111-911 220111-910
Bromoacetyl bromide ≥98%
Bromoacetyl bromide
Formula: BrCH₂COOBr
MW: 201.85 g/mol
Boiling Pt: 148 °C (1013 hPa)
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000115
CAS Number: 598-21-0
EINECS: 209-923-5
UN: 2513
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR