3'-Bromoacetophenone ≥99%

Warning

Synonyms: m-Bromoacetophenone

B56358-100G B56358-25G
220111-904EA 110.92 USD
220111-904 220111-905
3'-Bromoacetophenone ≥99%
3'-Bromoacetophenone
Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 255 °C (1013 hPa)
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000083
CAS Number: 2142-63-4
EINECS: 218-396-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR