Benzhydryl alcohol ≥99%

Synonyms: Diphenylcarbinol, Benzhydrol, Diphenylmethanol

B4856-5G B4856-500G B4856-100G
220111-827EA 35.32 USD
220111-827 220111-826 220111-825
Benzhydryl alcohol ≥99%
Benzhydryl alcohol
Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 297 °C (1013 hPa)
Melting Pt: 63…64 °C
Density: 1.054 g/cm³ (80 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004488
CAS Number: 91-01-0
EINECS: 202-033-8
Merck Index: 12,01121

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR