D-(+)-Biotin

B4501-500MG B4501-100MG B4501-1G B4501-10G B4501-25G B4501-5G
220111-750EA 89.6 USD
220111-750 220111-746 220111-748 220111-747 220111-749 220111-751
D-(+)-Biotin
D-(+)-Biotin
Formula: C₁₀H₁₆N₂O₃S
Melting Pt: 232…233 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005541
CAS Number: 58-85-5
EINECS: 200-399-3
Merck Index: 13,01231

Order Now

Specification Test Results

Assay 99% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR