Pyromellitic acid ≥96%

Warning

Synonyms: 1,2,4,5-Benzenetetracarboxylic acid, Benzene-1,2,4,5-tetracarboxylic acid

B4007-100G-A B4007-500G-A
220111-660EA 49.75 USD
220111-660 220111-661
Pyromellitic acid ≥96%
Pyromellitic acid
Formula: C₁₀H₆O₈
MW: 254.15 g/mol
Boiling Pt: 516 °C (1013 hPa)
Melting Pt: 272 °C
Density: 1.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002471
CAS Number: 89-05-4
EINECS: 201-879-5
Merck Index: 12,08188

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR