4-Phenylbenzoic acid ≥95%

Warning

Synonyms: Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid

B34729-25G B34729-100G
220111-543EA 55.46 USD
220111-543 220111-542
4-Phenylbenzoic acid ≥95%
4-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 373 °C (1013 hPa)
Melting Pt: 220 °C
Density: 1.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002553
CAS Number: 92-92-2
EINECS: 202-203-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR