(Benzyl)triphenylphosphonium chloride

Danger

Synonyms: Benzyltriphenylphosphonium chloride

B32807-100G B32807-25G
220111-474EA 98.75 USD
220111-474 220111-475
(Benzyl)triphenylphosphonium chloride
(Benzyl)triphenylphosphonium chloride
Formula: C₂₅H₂₂ClP
MW: 388.88 g/mol
Melting Pt: 345…347 °C
Density: 1.18 g/cm³ (20 °C)
MDL Number: MFCD00011913
CAS Number: 1100-88-5
EINECS: 214-154-3
UN: 3464
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR