5-Benzyloxytryptamine hydrochloride

Synonyms: 2-[5-(Benzyloxy)-1H-indol-3-yl]ethanamine hydrochloride, 5-(phenylmethoxy)indole-3-ethanamine monohydrochloride

B28656-1G B28656-250MG
220110-860EA 170.15 USD
220110-860 220110-861
5-Benzyloxytryptamine hydrochloride
5-Benzyloxytryptamine hydrochloride
Formula: C₁₇H₁₉ClN₂O
MDL Number: MFCD00012685
CAS Number: 52055-23-9
EINECS: 257-626-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR