5-Benzyloxyindole

Synonyms: 5-Benzyloxy-1H-indole, 5-(Benzyloxy)-1H-indole

B27803-5G B27803-25G
220110-855EA 34.89 USD
220110-855 220110-854
5-Benzyloxyindole
5-Benzyloxyindole
Formula: C₁₅H₁₃NO
MW: 223.27 g/mol
Boiling Pt: 165 °C (0.1 mmHg)
Melting Pt: 107 °C
MDL Number: MFCD00005676
CAS Number: 1215-59-4
EINECS: 214-930-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR