3-(Benzyloxy)benzaldehyde ≥98%

Synonyms: 3-Benzyloxybenzaldehyde

B27005-25G B27005-100G B27005-5G
220110-847EA 172.68 USD
220110-847 220110-846 220110-848
3-(Benzyloxy)benzaldehyde ≥98%
3-(Benzyloxy)benzaldehyde
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 119…120 °C (0.02 mmHg)
Melting Pt: 147…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00003367
CAS Number: 1700-37-4
EINECS: 216-932-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR