Benzyl(methyl)amine ≥97%

Warning

Synonyms: N-Benzylmethylamine, N-Methyl-1-phenylmethanamine, N-Methylbenzylamine

B25606-100ML B25606-5ML B25606-500ML
220111-296EA 33.28 USD
220111-296 220111-298 220111-297
Benzyl(methyl)amine ≥97%
Benzyl(methyl)amine
Formula: C₆H₅CH₂NHCH₃
MW: 121.18 g/mol
Boiling Pt: ∼184 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00008289
CAS Number: 103-67-3
EINECS: 203-133-4
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=067337f4cbd3ebd20fe522a5bac5c9f29cb10e6a&vid=fab9de39bcd5ba3758c9402492897f6aed55e8ee&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR