Benzyl mercaptan ≥99%

Synonyms: Toluene-α-thiol, α-Toluenethiol

B25401-100G B25401-5G
220111-240EA 57.94 USD
220111-240 220111-241
Benzyl mercaptan ≥99%
Benzyl mercaptan
Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 29 °C
Density: 1.055 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00004867
CAS Number: 100-53-8
EINECS: 202-862-5
UN: 2810
ADR: 6.1,II
Merck Index: 12,09456

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR