Barium hydroxide octahydrate ≥98%

Danger

B2507-1KG B2507-100G B2507-500G
220111-220EA 62.77 USD
220111-220 220111-219 220111-221
Barium hydroxide octahydrate ≥98%
Barium hydroxide octahydrate
Formula: Ba(OH)₂·8H₂O
MW: 315.47 g/mol
Boiling Pt: 780 °C (1013 hPa)
Melting Pt: 78 °C
Density: 2.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149152
CAS Number: 12230-71-6
EINECS: 241-234-5
UN: 3262
ADR: 8,III
Merck Index: 13,00980

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR