Benzyl bromide ≥98%, Reagent Grade

Warning

Synonyms: alpha-Bromotoluene

B17905-100G B17905-25G B17905-500G
220111-126EA 73.55 USD
220111-126 220111-127 220111-128
Benzyl bromide ≥98%, Reagent Grade
Benzyl bromide
Formula: (C₆H₅)CH₂Br
MW: 171.04 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.438 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000172
CAS Number: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1,II
Merck Index: 13,01130

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR