3-Benzoylpropionic acid ≥99%

Warning

Synonyms: 4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid

B13802-100G B13802-25G
220111-058EA 381.23 USD
220111-058 220111-059
3-Benzoylpropionic acid ≥99%
3-Benzoylpropionic acid
Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 114…117 °C
MDL Number: MFCD00002792
CAS Number: 2051-95-8
EINECS: 218-135-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR