Benzhydrazide ≥98%

Synonyms: Benzoylhydrazine, Benzoic acid hydrazide, Benzohydrazide, Benzoic hydrazide

B13071-25G B13071-100G
220111-026EA 60.31 USD
220111-026 220111-025
Benzhydrazide ≥98%
Benzhydrazide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 112 - 114 °C
MDL Number: MFCD00007596
CAS Number: 613-94-5
EINECS: 210-363-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR