1-Phenyl-1,3-butandione ≥99%

Synonyms: 1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione

B11907-50G B11907-10G B11907-250G
220110-575EA 174.46 USD
220110-575 220110-573 220110-574
1-Phenyl-1,3-butandione ≥99%
1-Phenyl-1,3-butandione
Formula: C₆H₅C(O)CH₂C(O)CH₃
MW: 162.19 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 57…59 °C
Density: 1.0899 g/cm³ (60 °C)
MDL Number: MFCD00008786
CAS Number: 93-91-4
EINECS: 202-286-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR