2-Butyne-1,4-diol ≥99%

Danger

Synonyms: 1,4-Dihydroxy-2-butyne, But-2-yne-1,4-diol

B103209-1KG B103209-100G B103209-500G
220110-491EA 72.26 USD
220110-491 220110-972 220110-492
2-Butyne-1,4-diol ≥99%
2-Butyne-1,4-diol
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 55 - 58 °C
Density: 1.1001 g/cm³ (20 °C)
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00002915
CAS Number: 110-65-6
EINECS: 203-788-6
UN: 2716
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR