2,3-Butanedione monoxime ≥98%

Synonyms: Diacetyl monoxime

B0753-100G B0753-25G B0753-1KG B0753-500G
220110-931EA 152.58 USD
220110-931 220110-933 220110-932 220110-934
2,3-Butanedione monoxime ≥98%
2,3-Butanedione monoxime
Formula: C₄H₇NO₂
MW: 101.11 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002116
CAS Number: 57-71-6
EINECS: 200-348-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR