Barium chloride dihydrate ≥99%

Danger

Synonyms: Barium dichloride

B0750-500G B0750-100G B0750-1KG
220110-929EA 47.2 USD
220110-929 220110-818 220110-819
Barium chloride dihydrate ≥99%
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3.86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR