β-Alanine

Synonyms: H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid

A9920-100G A9920-10MG
220105-711EA 37 USD
220105-711 220105-712
β-Alanine
β-Alanine
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5
Merck Index: 13,00204

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR