7-Amino-4-methylcoumarin

Synonyms: 7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC

A9891-250MG A9891-1G A9891-500MG
220105-048EA 189.23 USD
220105-048 220105-047 220105-049
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 226 °C
Density: 1.377 g/cm³ (20 °C)
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR