2-Aminobenzaldehyde

Warning

A9628-100MG A9628-1G A9628-500MG
220105-599EA 112 USD
220105-599 220105-600 220105-601
2-Aminobenzaldehyde
2-Aminobenzaldehyde
Formula: C₇H₇NO
MW: 121.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00007709
CAS Number: 529-23-7
EINECS: 208-454-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR