9,10-Anthraquinone

Synonyms: Anthracene-9,10-dione, Anthraquinone

A90004-250G A90004-50G
220105-264EA 86.8 USD
220105-264 220105-265
9,10-Anthraquinone
9,10-Anthraquinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 165 - 169 °C
Density: 1.419 to 1.438 g/cm³ (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR