9-Anthracenecarboxylic acid ≥99%

Synonyms: 9-Anthroic acid, Anthracene-9-carboxylic acid, 9-Anthracenecarboxylic acid, Anthracene-9-carboxylic acid, 9-Carboxyanthracene

A89405-25G A89405-5G
220105-231EA 226.15 USD
220105-231 220105-232
9-Anthracenecarboxylic acid ≥99%
9-Anthracenecarboxylic acid
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 411 °C (1013 hPa)
Melting Pt: 214…217 °C
Density: 1.397 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001257
CAS Number: 723-62-6
EINECS: 211-964-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR