p-Anisidine ≥99%

Danger

Synonyms: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline

A88255-1KG A88255-100G A88255-5G A88255-250G
220105-189EA 166.92 USD
220105-189 220105-188 220105-191 220105-190
p-Anisidine ≥99%
p-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 55…58 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00007864
CAS Number: 104-94-9
EINECS: 203-254-2
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR