m-Anisidine ≥97%

Danger

Synonyms: 3-Aminoanisole, 3-Methoxyaniline, 3-Anisidine

A88204-1KG A88204-250G A88204-5G A88204-100G
220105-185EA 586.92 USD
220105-185 220105-186 220105-187 220105-184
m-Anisidine ≥97%
m-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007783
CAS Number: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR